Learn More
R&L Enterprises™ Bossheads
For securely keeping clamps in place on laboratory scaffolding. R&L Enterprises™ Bossheads come with offset jaws for greater strength.
Brand:  R&L Enterprises™ Z0204
Product Code. 11765386
  • 11.30 € / Each
Estimated Shipment: 02-07-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Aluminum alloy with clamping screws
  • For use with rods up to 16mm in diameter
Specifications
Specifications
Bosshead Clamp
Alloy
with Blue Plastic headed nickel plated steel screws
For use with rods up to 16mm diameter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
R&L Enterprises™ Bossheads > For Use With: For use with rods up to 16mm diameter