missing translation for 'onlineSavingsMsg'
Learn More

Manutan Alloy rope hook with catch, medium duty, 13mm diameter

Brand:  Manutan A339944

Encompass_Preferred

Product Code. 12861399

  • 16.19 € / Each
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Complete with catch. Available with standard or with swivel hook.
  • Standard
  • Rope diameter: 13mm
  • WLL: 2000kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers