missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manutan Alloy rope hook with catch, medium duty, 16mm diameter
Brand: Manutan A340003
This item is not returnable.
View return policy
Description
Complete with catch. Available with standard or with swivel hook.- Standard
- Rope diameter: 16mm
- WLL: 3000kg