missing translation for 'onlineSavingsMsg'
Learn More

Manutan Alloy swivel rope hook with catch, light duty, 3mm diameter

Brand:  Manutan A339947

Encompass_Preferred

Product Code. 12888728

  • 20.75 € / Each
Estimated Shipment: 23-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Complete with catch. Available with standard or with swivel hook.
  • Swivel
  • WLL: 100kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers