Learn More
Allyl chloroformate, 97%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 2937-50-0 |
---|---|
Molecular Formula | C4H5ClO2 |
Molecular Weight (g/mol) | 120.54 |
MDL Number | MFCD00000648 |
InChI Key | CAEWJEXPFKNBQL-UHFFFAOYSA-N |
Synonym | allyl chloroformate, chloroformic acid allyl ester, allyl chlorocarbonate, allylchloroformate, 2-propenyl chloroformate, carbonochloridic acid, 2-propenyl ester, formic acid, chloro-, allyl ester, hsdb 621, allylester kyseliny chlormravenci, allylester kyseliny chlormravenci czech |
PubChem CID | 18052 |
IUPAC Name | prop-2-enyl carbonochloridate |
SMILES | C=CCOC(=O)Cl |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Coupling reagentChemical Identifiers
2937-50-0 | |
120.54 | |
CAEWJEXPFKNBQL-UHFFFAOYSA-N | |
18052 | |
C=CCOC(=O)Cl |
C4H5ClO2 | |
MFCD00000648 | |
allyl chloroformate, chloroformic acid allyl ester, allyl chlorocarbonate, allylchloroformate, 2-propenyl chloroformate, carbonochloridic acid, 2-propenyl ester, formic acid, chloro-, allyl ester, hsdb 621, allylester kyseliny chlormravenci, allylester kyseliny chlormravenci czech | |
prop-2-enyl carbonochloridate |
Specifications
2937-50-0 | |
100.0 | |
1.1300g/mL | |
31°C | |
96% min. (GC) | |
ClCO2CH2CH=CH2 | |
MFCD00000648 | |
12,15; 13,9 | |
allyl chloroformate, chloroformic acid allyl ester, allyl chlorocarbonate, allylchloroformate, 2-propenyl chloroformate, carbonochloridic acid, 2-propenyl ester, formic acid, chloro-, allyl ester, hsdb 621, allylester kyseliny chlormravenci, allylester kyseliny chlormravenci czech | |
CAEWJEXPFKNBQL-UHFFFAOYSA-N | |
prop-2-enyl carbonochloridate | |
18052 | |
97% | |
Allyl chloroformate |
96.0 | |
Brown-Red to Yellow | |
109.0°C to 110.0°C | |
Authentic | |
C4H5ClO2 | |
1.4213 to 1.4233 | |
03, 12 | |
1.13 | |
Solubility in water: decomposes | |
C=CCOC(=O)Cl | |
120.54 | |
120.54 | |
Liquid |
Safety and Handling
GHS H Statement
Fatal if inhaled.
Causes severe skin burns and eye damage.
Toxic if swallowed.
Flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 220-916-6
- Allyl chloroformate
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 3
- Acute toxicity Category 1
- Skin corrosion/irritation Category 1 B
- Serious eye damage/eye irritation Category 1
Hazard Statement
- H226-Flammable liquid and vapour.
- H301-Toxic if swallowed.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P310-Immediately call a POISON CENTER/doctor/.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.