missing translation for 'onlineSavingsMsg'
Learn More
Allyl ethyl sulfide, 97%, Thermo Scientific Chemicals Product Code.: 11393396

Allyl ethyl sulfide, 97%, Thermo Scientific Chemicals

Product Code. 11393396
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11393396

Brand: Thermo Scientific Alfa Aesar L12847.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5296-62-8
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.20
MDL Number MFCD00026979
InChI Key NOJXPGXFDASWEI-UHFFFAOYSA-N
Synonym allyl ethyl sulfide, 4-thia-1-hexene, 1-propene-1-one, 3-ethylthio, allylethylsulfide, allyl ethylsulfide, allylethyl sulfide, allyl ethyl sulphide, 3-ethylthio-1-propene, acmc-1ao1o
PubChem CID 123229
IUPAC Name 3-ethylsulfanylprop-1-ene
SMILES CCSCC=C

Specifications

Boiling Point 115°C to 116°C
Flash Point 18°C (64°F)
Odor Unpleasant
Linear Formula CH3CH2SCH2CH=CH2
Quantity 1 g
UN Number UN1993
Beilstein 1736876
Formula Weight 102.19
Percent Purity 97%
Chemical Name or Material Allyl ethyl sulfide

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10116392 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberL12847.06,L12847.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Allyl ethyl sulfide, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.