Learn More
Allyl iodide, 97%, stabilized, Thermo Scientific Chemicals
84.42 € - 691.00 €
Chemical Identifiers
CAS | 556-56-9 |
---|---|
Molecular Formula | C3H5I |
Molecular Weight (g/mol) | 167.98 |
MDL Number | MFCD00001094 |
InChI Key | HFEHLDPGIKPNKL-UHFFFAOYSA-N |
Synonym | allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene |
PubChem CID | 11166 |
IUPAC Name | 3-iodoprop-1-ene |
SMILES | ICC=C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10236370
|
Thermo Scientific Acros
218540250 |
25 mL | Glass bottle |
84.42 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10216420
|
Thermo Scientific Acros
218541000 |
100 mL | Glass bottle |
262.31 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11954501
|
Thermo Scientific Acros
218545000 |
500 mL | Glass Bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
556-56-9 | |
167.98 | |
HFEHLDPGIKPNKL-UHFFFAOYSA-N | |
11166 | |
ICC=C |
C3H5I | |
MFCD00001094 | |
allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene | |
3-iodoprop-1-ene |
Specifications
556-56-9 | |
Brown-Red or Orange-Red to Yellow | |
102.0°C to 103.0°C | |
Authentic | |
C3H5I | |
1.5530 to 1.5550 | |
01,202 | |
1.83 | |
Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform and ether | |
ICC=C | |
Copper | |
11166 | |
97% | |
Allyl iodide |
-99.0°C | |
1.8300g/mL | |
18°C | |
96% min. (GC) | |
H2C=CHCH2I | |
MFCD00001094 | |
15,285 | |
allyl iodide, 1-propene, 3-iodo, 3-iodopropene, 3-iodo-1-propene, 3-iodopropylene, propene, 3-iodo, allyliodide, unii-46830qoa4d, 3-iodo-propene, 3-iodanylprop-1-ene | |
HFEHLDPGIKPNKL-UHFFFAOYSA-N | |
3-iodoprop-1-ene | |
167.98 | |
167.98 | |
Liquid |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Fatal if swallowed.
Highly flammable liquid and vapor.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 209-130-4
- Allyl iodide, stabilized
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Serious eye damage/eye irritation Category 1
- Flammable liquid Category 2
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H300-Fatal if swallowed.
- H225-Highly flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P240-Ground/bond container and receiving equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.