missing translation for 'onlineSavingsMsg'
Learn More

Allyl isothiocyanate, 94%, stabilized with 0.01% alpha-tocopherol, Thermo Scientific Chemicals

Product Code. 10224130
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57-06-7
MDL Number MFCD00004822
InChI Key ZOJBYZNEUISWFT-UHFFFAOYSA-N
Synonym allyl isothiocyanate, mustard oil, redskin, allylsenevol, allylsenfoel, allyl mustard oil, oleum sinapis, oil of mustard, allylsevenolum, senfoel
PubChem CID 5971
ChEBI CHEBI:73224
IUPAC Name 3-isothiocyanatoprop-1-ene
SMILES C=CCN=C=S

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point -80.0°C
Color White
Density 1.0175g/mL
Boiling Point 151.0°C to 153.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 92% min. (GC)
Packaging Glass bottle
Molecular Formula C4H5NS
Linear Formula H2C=CHCH2NCS
Refractive Index 1.5290 to 1.5310
Quantity 5 g
Beilstein 04, 214
Merck Index 15, 286
Specific Gravity 1.0175
Solubility Information Solubility in water: 2g/L water (20°C). Other solubilities: ethanol of 65 vol%- 1:20 v/v,diethylphthalate - in all proportions,paraffin oil 65 cp- in all proportions,1,2-propanediol - 1:5 v/v,soluble in ether,acetone,benzene,cs2
Stabilizer 0.01% alpha-tocopherol
Molecular Weight (g/mol) 99.15
Formula Weight 99.15
Percent Purity 94%
Physical Form Fine Crystalline Powder
Chemical Name or Material Allyl isothiocyanate, 94%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allyl isothiocyanate, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H301-Toxic if swallowed.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'