missing translation for 'onlineSavingsMsg'
Learn More

Allyl sulfide, 98%, Thermo Scientific Chemicals

Product Code. 10127523
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-88-1
Molecular Formula C6H10S
Molecular Weight (g/mol) 114.21
MDL Number MFCD00008658
InChI Key UBJVUCKUDDKUJF-UHFFFAOYSA-N
Synonym diallyl sulfide, allyl sulfide, oil garlic, diallylsulfide, thioallyl ether, 1-propene, 3,3'-thiobis, allyl monosulfide, diallyl thioether, diallyl monosulfide, diallyl sulphide
PubChem CID 11617
ChEBI CHEBI:4489
IUPAC Name 3-prop-2-enylsulfanylprop-1-ene
SMILES C=CCSCC=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -83.0°C
Color Colorless
Density 0.8870g/mL
Boiling Point 138.0°C
Flash Point 46°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula (H2C=CHCH2)2S
Refractive Index 1.4879 to 1.4899
Quantity 100 mL
Beilstein 01, 440
Merck Index 15, 288
Specific Gravity 0.887
Solubility Information Solubility in water: insoluble. Other solubilities: miscible with alcohol,chloroform,ether and,carbon tetrachloride
Formula Weight 114.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Allyl sulfide
Show More Show Less
compliance-icons
Product Identifier
  • Allyl sulfide
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'