missing translation for 'onlineSavingsMsg'
Learn More
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals Product Code.: 10081544

Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals

Product Code. 10081544
100 mL, AcroSeal™ Glass bottle
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

Product Code. 10081544

Brand: Thermo Scientific Acros 209531000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
This item is packed in AcroSeal™, the industry-leading packaging for air and moisture sensitive reagents.
Find out more about AcroSeal™ packaging.
 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1730-25-2
Molecular Formula C3H5BrMg
Molecular Weight (g/mol) 145.28
MDL Number MFCD00000044
InChI Key FEMBXICCJNZMMC-UHFFFAOYSA-M
Synonym allylmagnesium bromide, allyl magnesium bromide, allylmagnesium bromide solution, sjpdabpprujp@, allylmagnesium bromide solution, 1.0 m in diethyl ether, allylmagnesiumbromid, allylmagnesiumbromide, allyl magnesiumbromide, allyl-magnesium bromide, 2-propenylmagnesium bromide, grignard reagentShow MoreShow Less
PubChem CID 74413
IUPAC Name magnesium;prop-1-ene;bromide
SMILES Br[Mg]CC=C

Specifications

CAS Min % 15.37, 81.22
CAS Max % 18.78, 84.64
Color Gray or Green-Yellow
Density 0.8510g/mL
Flash Point −40°C
Packaging AcroSeal™ Glass bottle
Linear Formula H2C=CHCH2MgBr
Quantity 100 mL
UN Number 3399
Specific Gravity 0.851
Formula Weight 145.28
Physical Form Solution
Chemical Name or Material Allylmagnesium bromide, 1M solution in diethyl ether
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0463186 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number209538000,209531000
Lot NumberA0463911 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number209538000,209531000
Lot NumberA0463911 Certificate TypeCertificate of Analysis Date28/06/2024 Catalog Number209538000,209531000
Lot NumberA0463186 Certificate TypeCertificate of Analysis Date24/05/2024 Catalog Number209538000,209531000
Lot NumberA0462464 Certificate TypeCertificate of Analysis Date03/05/2024 Catalog Number209538000,209531000
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Allylmagnesium bromide, 1M solution in diethylether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 1
  • Pyrophoric liquid Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity Category 3
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H336-May cause drowsiness or dizziness.
  • H302-Harmful if swallowed.
  • H250-Catches fire spontaneously if exposed to air.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • H224-Extremely flammable liquid and vapour.
  • EUH014-Reacts violently with water.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Allylmagnesium bromide, 1M solution in diethyl ether, AcroSeal™, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.