missing translation for 'onlineSavingsMsg'
Learn More

Allylthiourea, 98%

Product Code. 10133541 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy

Product Code. 10133541

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-57-9 | C4H8N2S | 116.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-57-9
MDL Number MFCD00004940
InChI Key HTKFORQRBXIQHD-UHFFFAOYSA-N
Synonym allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
PubChem CID 1549517
ChEBI CHEBI:74079
IUPAC Name prop-2-enylthiourea
SMILES C=CCNC(=S)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70.0°C to 78.0°C
Color White to Beige
Loss on Drying 1.5% max. (50°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C4H8N2S
Linear Formula H2C=CHCH2NHCSNH2
Quantity 25 g
Beilstein 04, 211
Merck Index 15, 9515
Solubility Information Solubility in water: 67g/L (20°C). Other solubilities: 1370g/L methanol (20°C)- insoluble in benzene,soluble in ether,solutions of borax,benzoates,and urethane,slightly soluble in 70% alcohol
Molecular Weight (g/mol) 116.18
Formula Weight 116.18
Percent Purity 98%
Physical Form Crystals, Powder, Granules or Chunks
Chemical Name or Material Allylthiourea, 98%
Show More Show Less
compliance-icons
Product Identifier
  • Allylthiourea
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.