missing translation for 'onlineSavingsMsg'
Learn More

Allyltriethoxysilane, 97%, Thermo Scientific Chemicals

Product Code. 11326886
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Allyltriethoxysilane is used as chemical additives and intermediates. It is used as general catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltriethoxysilane is used as chemical additives and intermediates. It is used as general catalyst.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from oxidizing agents. Incompatible with moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-04-1
MDL Number MFCD00009064

Specifications

Boiling Point 176°C to 177°C
Quantity 5 g
Molecular Formula C9H20O3Si
Linear Formula CH2=CHCH2Si(OCH2CH3)3
UN Number UN1993
Beilstein 1772306
Flash Point 21°C (69°F)
Synonym allyltriethoxysilane, silane, triethoxy-2-propenyl, triethoxyallylsilane, silane, allyltriethoxy, allytriethoxysilane, triethoxy prop-2-en-1-yl silane, silane, triethoxy-2-propen-1-yl, allyl triethoxysilane, acmc-2097bb
Solubility Information Hydrolyzes in water.
InChI Key UMFJXASDGBJDEB-UHFFFAOYSA-N
SMILES CCO[Si](CC=C)(OCC)OCC
IUPAC Name triethoxy(prop-2-enyl)silane
Molecular Weight (g/mol) 204.34
PubChem CID 75692
Formula Weight 204.34
Percent Purity 97%
Refractive Index 1.407
Sensitivity Moisture sensitive
Density 0.953
Chemical Name or Material Allyltriethoxysilane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'