Learn More
3M™ Almond Protein Rapid Kit L25ALM
Intended for screening for the presence of almond proteins in clean-in-place (CIP) final rinse water, environmental swab samples, food ingredients and processed food products.
Brand:  3M™ 7100151319
Encompass
Product Code. 17233794
  • 486.99 € / 25 tests
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Results visible at 11 ± 1 minute after application of the sample.
  • Rapid, qualitative immunochromatographic lateral flow test cartridges designed for the detection of allergenic food proteins.
  • Designed to detect both processed and unprocessed allergen proteins.
  • Test can accommodate environmental swab, CIP rinse water and food samples.
Specifications
Specifications
Microbiology Media
Allergen
25/Cs.
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Almond Protein Rapid Kit
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, glycerol, urea, sodium chloride