missing translation for 'onlineSavingsMsg'
Learn More
alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%, Thermo Scientific Chemicals Product Code.: 10307120

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%, Thermo Scientific Chemicals

Product Code. 10307120
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 10307120

Brand: Thermo Scientific Acros 137971000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13209-15-9
MDL Number MFCD00000131
InChI Key LNAOKZKISWEZNY-UHFFFAOYSA-N
Synonym 1,2-bis dibromomethyl benzene, alpha,alpha,alpha',alpha'-tetrabromo-o-xylene, benzene, 1,2-bis dibromomethyl, unii-3wg2h68vx4, o-xylylene tetrabromide, alpha,alpha,alpha,alpha-tetrabromo-o-xylene, a,a,a',a'-tetrabromo-o-xylene, o-xylene, alpha,alpha,alpha',alpha'-tetrabromo, alpha,alpha,beta,beta-tetrabromo-o-xylene, acmc-209boiShow MoreShow Less
PubChem CID 83234
IUPAC Name 1,2-bis(dibromomethyl)benzene
SMILES C1=CC=C(C(=C1)C(Br)Br)C(Br)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 114°C to 118°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Molecular Formula C8H6Br4
Linear Formula C6H4(CHBr2)2
Quantity 100 g
Beilstein 05,367
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 421.75
Formula Weight 421.75
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material α,α,α',α'-Tetrabromo-o-xylene, 97%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465387 Certificate TypeCertificate of Analysis Date21/08/2024 Catalog Number137971000,137970250
Lot NumberA0413416 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number137971000,137970250
Lot NumberA0433983 Certificate TypeCertificate of Analysis Date21/09/2023 Catalog Number137971000,137970250
Lot NumberA0405064 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number137971000,137970250
Lot NumberA0395150 Certificate TypeCertificate of Analysis Date22/03/2021 Catalog Number137971000,137970250
compliance-icons compliance-icons compliance-icons
Product Identifier
  • a,a,a',a'-Tetrabromo-o-xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.