missing translation for 'onlineSavingsMsg'
Learn More

alpha,alpha,alpha',alpha'-Tetrabromo-o-xylene, 97%

Product Code. p-195432 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10072843

Brand: Thermo Scientific Acros 137970250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13209-15-9 | C8H6Br4 | 421.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13209-15-9
MDL Number MFCD00000131
InChI Key LNAOKZKISWEZNY-UHFFFAOYSA-N
Synonym 1,2-bis dibromomethyl benzene, alpha,alpha,alpha',alpha'-tetrabromo-o-xylene, benzene, 1,2-bis dibromomethyl, unii-3wg2h68vx4, o-xylylene tetrabromide, alpha,alpha,alpha,alpha-tetrabromo-o-xylene, a,a,a',a'-tetrabromo-o-xylene, o-xylene, alpha,alpha,alpha',alpha'-tetrabromo, alpha,alpha,beta,beta-tetrabromo-o-xylene, acmc-209boi
PubChem CID 83234
IUPAC Name 1,2-bis(dibromomethyl)benzene
SMILES C1=CC=C(C(=C1)C(Br)Br)C(Br)Br

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 114°C to 118°C
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Plastic bottle
Molecular Formula C8H6Br4
Linear Formula C6H4(CHBr2)2
Quantity 25 g
Beilstein 05,367
Solubility Information Solubility in water: insoluble
Molecular Weight (g/mol) 421.75
Formula Weight 421.75
Percent Purity 97%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material α,α,α',α'-Tetrabromo-o-xylene, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • a,a,a',a'-Tetrabromo-o-xylene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.