missing translation for 'onlineSavingsMsg'
Learn More

alpha-Amino-gamma-butyrolactone hydrobromide, 99%, Thermo Scientific Chemicals

Product Code. 11300496
Click to view available options
Quantity:
2.5 g
10 g
50 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6305-38-0
Molecular Formula C4H7NO2·BrH
Molecular Weight (g/mol) 182.02
MDL Number MFCD00012723
InChI Key MKLNTBLOABOJFZ-UHFFFAOYSA-N
Synonym alpha-amino-gamma-butyrolactone hydrobromide, 3-aminooxolan-2-one hydrobromide, hsls hydrobromide, 3-aminodihydrofuran-2 3h-one hydrobromide, 2-amino-4-butyrolactone, 2 3h-furanone, hydrobromide, 2 3h-furanone, 3-aminodihydro-, hydrobromide, 2 3h-furanone,3-aminodihydro-, hydrobromide 1:1, hydrobromide alpha-amino-gamma-butyrolactone, l-homoserine lactone hbr
PubChem CID 73508
IUPAC Name 3-aminooxolan-2-one;hydrobromide
SMILES C1COC(=O)C1N.Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 221.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (Argentometry)
Packaging Glass bottle
Quantity 2.5 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in ethanol,acetone
Formula Weight 182.02
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material α-Amino-γ-butyrolactone hydrobromide, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'