missing translation for 'onlineSavingsMsg'
Learn More

Skytec™ Alpha Sentinel Premium Reusable Full Face Mask

Alpha Sentinel Full Face Masks are ergonomically designed for optimum protection, comfort and PPE compatibility.

243.85 €

Specifications

Certifications/Compliance CE , UKCA , EN 136:1998 Class 2
Certifications (EN 136) Class 2
Color Black
Filter Type HEPA, Combination Organic Vapor/Acid Gas/HEPA, Multi-Gas/HEPA
Product Type Full Facepiece Respirator
View More Specs

Products 3
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
30242954
View Documents
Skytec™
ASRAS0002BD
Small
243.85 €
Each
Please to purchase this item. Need a web account? Register with us today!
30242980
View Documents
Skytec™
ASRAS0002BF
Medium
243.85 €
Each
Please to purchase this item. Need a web account? Register with us today!
30242985
View Documents
Skytec™
ASRAS0002BH
Large
243.85 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • The five-point adjustable head harness removes the feeling of a downward pull, ensuring a balanced, comfortable fit
  • Well-balanced, lightweight (600 g)
  • Available in three sizes for a more tailored fit, increasing respirator/fit factor
  • Raised visor bridge profile protects the visor from scratches when not in use
  • Visor and filter position provides panoramic field of view without any obstructions
  • Low profile shape provides exceptional compatibility with other PPE
  • The Alpha Sentinel filters are fitted with a simple connection which easily clicks into position
  • The large exhalation valve reduces heat and moisture build-up, providing low breathing resistance
  • Easily cleaned and maintained, with spare parts available
  • Tested to EN136:1998 Class 2, ensuring greater robustness and higher flame retardancy
  • Supplied with user seal check covers, which double as particulate filter holder for combination filter
  • Optical insert available
  • Quantitative face fit adapter available
  • The harness is designed to prevent over-rotation and tangling
Specifications

Specifications

CE , UKCA , EN 136:1998 Class 2
Black
Full Facepiece Respirator
Thermoplastic Elastomer
Class 2
HEPA, Combination Organic Vapor/Acid Gas/HEPA, Multi-Gas/HEPA
Yes
Alpha Sentinel Full Face Mask
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.