missing translation for 'onlineSavingsMsg'
Learn More
alpha-Terpinene, 90+%, Thermo Scientific Chemicals Product Code.: 15485169

alpha-Terpinene, 90+%, Thermo Scientific Chemicals

Product Code. 15485169
100 mL
Click to view available options
Quantity:
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 15485169

Brand: Thermo Scientific Alfa Aesar J62871.AE

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It can be used as a chemical fragrance essential oil and spices. It is used as a flavoring and fragrance chemical used in the personal care and cosmetic and food industries. Also used in the pharmaceutical, electronics, food processing, semi-conductor manufacturing industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as a chemical fragrance essential oil and spices. It is used as a flavoring and fragrance chemical used in the personal care and cosmetic and food industries. Also used in the pharmaceutical, electronics, food processing, semi-conductor manufacturing industries.

Solubility
Soluble in ether, alcohol, and ethanol (100 mg/ml). Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Recommended storage temperature: -20°C. Store under inert gas. Air sensitive. Keep away from strong acids, strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-86-5
Molecular Formula C10H16
Molecular Weight (g/mol) 136.24
MDL Number MFCD00001534
InChI Key YHQGMYUVUMAZJR-UHFFFAOYSA-N
Synonym alpha-terpinene, p-mentha-1,3-diene, terpilene, 1-isopropyl-4-methyl-1,3-cyclohexadiene, 1,3-cyclohexadiene, 1-methyl-4-1-methylethyl, alpha-terpinen, 1-methyl-4-1-methylethyl-1,3-cyclohexadiene, 1-methyl-4-isopropylcyclohexadiene-1,3, .alpha.-terpinen, .alpha.-terpinene
PubChem CID 7462
ChEBI CHEBI:10334
IUPAC Name 1-methyl-4-propan-2-ylcyclohexa-1,3-diene
SMILES CC(C)C1=CC=C(C)CC1

Specifications

Color Colorless
Density 0.837
Boiling Point 173°C to 175°C
Flash Point 50°C (122°F)
Refractive Index 1.478
Quantity 100 mL
UN Number UN2319
Beilstein 1853379
Merck Index 14,9170
Solubility Information Soluble in ether,alcohol,and ethanol (100mg/ml). Insoluble in water.
Formula Weight 136.23
Percent Purity ≥90%
Physical Form Liquid
Chemical Name or Material alpha-Terpinene
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
alpha-Terpinene, 90+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.