missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Texwipe™ AlphaLite™ ll Wipers
Economical lightweight knitted wiper
Brand: Texwipe™ TX1091
83.39 EUR valid until 2024-12-31
Use promo code "21676" to get your promotional price.
This item is not returnable.
View return policy
Description
- 100% synthetic fiber (continuous-filament knitted polyester)
- Solvent-safe Bag-Within-A-Bag™, amide and silicone free cleanroom packaging
- Low particulate and extractable levels
- Good absorbency, wiping efficiency and abrasion resistant
- Lot to lot traceability
- Permits rigorous wiping with minimal release of fibers and particles
- For use with flat ‘cloth holder’ mops like FILMOP
- For spill, general wiping and maintenance of work surfaces
- Cleaning of floors and walls
- Excellent for multiple applications
- Can be used with DIW and IPA for cleaning critical surfaces
Specifications
60 cm | |
25 cm | |
2 Bags x 75 Wipes, 4 Bags/Cs. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction