missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 1500 STANDARD Stitched Coveralls, Model 138, White

Product Code. 18070140
Click to view available options
Size:
4X-Large
6X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides comfortable protection from hazardous particulates in general industrial environments.

SUIT FEATURES

  • 3 piece hood
  • Elasticated hood, wrists, waist and ankles
  • 2-way front zipper with resealable storm flap
TRUSTED_SUSTAINABILITY

Specifications

Product Type Chemical Protective Clothing
Size 4X-Large
Gender Unisex
Material SMS Fabric
Color White
Disposable Reusable
Waist Style Elastic
Ankle Style Elastic
Cuff Style Elastic
Neck Style Hooded
Hood Type 3-panel, Elastic
Closure Type 2-way Front Zipper with Storm Flap
Seam Type Stitched
Sterility Non-sterile
Protection Class Category III risks I Type 5/6
Resistance Chemicals, Particle
Collar Type 3-piece elasticated hood
Certifications/Compliance (Europe) CE-nummer|EN ISO 13688:2013|EN ISO 13982-1:2004 + A1:2010|EN ISO 13982-1:2004 + A1:2010|EN ISO 13982-1:2004 + A1:2010|EN 13034:2005 + A1:2009|EN 1073-2:2002
Latex Free Yes
For Use With (Application) Inspection (picking, checking, and marking), Asbestos Related Work, Handling of powders, General maintenance
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.