missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 1800 COMFORT Bound - Model 195
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
- 3 piece hood
- Elasticated hood, wrists, waist and ankles
- 2-way front zipper with resealable storm flap
- Finger Loops

Specifications
Specifications
Size | X-Large |
Material | Microporous Non-woven Polypropylene and Polyethylene Laminate |
Color | White, Blue |
Seam Type | Bound |
Coating | Anti-Static Treatment (Back, Underarm and Hood) |
Certifications/Compliance (Europe) | EN 374:2003, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2008, US Ergo Certification |
For Use With (Application) | Clean Room Cleaning and Preparing, Body Painting Including Surface Inspection, Production Line Support & Maintenance, Cleaning of Plant and Machines, Maintenance of Plant and Machines, Fibre Glass Resin Applications, Handling of Powders, General Purpose Auto Aftermarket |
Style | W18B195 |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell AlphaTec™ 1800 COMFORT Bound - Model 195 >
Spot an opportunity for improvement?Share a Content Correction