missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 STANDARD Apron
287.39 EUR valid until 2024-09-30
Use promo code "21675" to get your promotional price.
Use promo code "21675" to get your promotional price.
Click to view available options
Size:
Small
Universal
X-Large
Style:
W20B214
W20S213
Description
- Silicone Free
- Low Linting
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Size | X-Large |
For Use With (Application) | Clean Room Cleaning and Preparing, Production Line Support and Maintenance, Maintenance of Plant and Machines, Veterinary Services |
Style | W20B214 |
Material | Non-woven Microporous Polyethylene Laminate |
Color | White |
Seam Type | Bound |
Sleeve Style | Long Sleeve |
Certifications/Compliance | EN ISO 13688:2013, Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'