missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell™ AlphaTec™ 2000 STANDARD Bound
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Description
Provides both protection and comfort with exceptional liquid and particulate protection.
- Ideal for a wide range of industrial applications
- Low hazard liquid chemical repellence, particle protection, protection from pesticides and barrier to biological agents.
- Proven protective barrier to low concentration liquid chemicals, liquids, and particulate biological hazards
- Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
- Optimized body fit, improving wearer comfort and safety
- Anti-static, tested according to EN 1149-5
- Low linting to help reduce the risk of cross contamination in critical areas
- Silicone free, critical in spray painting applications

Specifications
Specifications
Product Type | Controlled Environments ESD Coveralls |
Size | 2X-Large |
Gender | Unisex |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White |
Disposable | Reusable |
Ankle Style | Elastic |
Cuff Style | Elastic |
Neck Style | Hooded |
Closure Type | Zipper |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ansell™ AlphaTec™ 2000 STANDARD Bound >
Spot an opportunity for improvement?Share a Content Correction