Learn More
Ansell AlphaTec™ 2000 STANDARD Lab Coat Bound Product Code.: 16854790
Add to basket
Ansell AlphaTec™ 2000 STANDARD Lab Coat Bound
Product Code. 16854790
X-Large
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
Product Code. 16854790
Brand: Ansell WH20B0023105
Add to basket
Add to basket
AlphaTec™ 2000 STANDARD Lab coat is designed to allow water vapour (perspiration) to escape from the suit yet will withstand saturation of liquid chemicals and filter 100% of particulates down to 0.01 microns in size.
Designed to allow perspiration to escape from the suit and withstand saturation of liquid chemicals.
  • Filter 100% of particulates down to 0.01 microns in size
  • Proven protective barrier to low concentration liquid chemicals, liquids, and particulate biological hazards
  • Comfortable to wear featuring moisture vapor permeable fabric to help reduce the risk of heat stress
  • Optimized fit, improving wearer comfort and safety
  • Low linting
TRUSTED_SUSTAINABILITY
Specifications
Product Type Controlled Environments Lab Coat
Size X-Large
Gender Unisex
Material Microporous Non-woven Polyethylene Laminate
Color White
Closure Type Zipper
Disposable Reusable
Resistance Fluids
Style Knee-length
Model 2000
Certifications/Compliance (Europe) CE CATEGORY III (Type PB (6)-B, EN 14126)
Autoclavable Not Autoclavable
Product Title
Ansell AlphaTec™ 2000 STANDARD Lab Coat Bound >