missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 2000 STANDARD lab coat Bound
343.44 EUR valid until 2024-12-31
Use promo code "22917" to get your promotional price.
Use promo code "22917" to get your promotional price.
Click to view available options
Style: W20B209
Description
- Proven protective barrier to low concentration liquid chemicals, liquid & particulate biological hazards
- Optimized head fit, improving wearer comfort and safety
- Low Linting
Specifications
Product Type | Controlled Environments Lab Coat |
Size | Small |
Material | Microporous Non-woven Polyethylene Laminate |
Color | White |
Certifications/Compliance | ASTM F1671 |
Certifications/Compliance (Europe) | Partial Body Protection Only, EN 13034:2005 + A1:2009, EN 14126:2003, EN ISO 13688:2013 |
For Use With (Application) | Cleanroom Cleaning and Preparing, Production Line Support & Maintenance, Maintenance of Plant and Machines, Veterinary Services |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'