Learn More
Ansell AlphaTec™ 2000 Ts PLUS
Product Code. 16874810
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
Style:
W20T103
W20T111
W20T122
Chemical protective suits that have a proven protective barrier to low concentration liquid chemicals, liquid and particulate biological hazards.
  • Comfortable to wear featuring moisture vapor permeable fabric (breathable) to help reduce the risk of heat stress
  • Optimized body fit, improving wearer comfort and safety
  • Low linting to help reduce the risk of cross contamination in critical areas
  • Silicone free, critical in spray painting applications
TRUSTED_SUSTAINABILITY
Specifications
Size 3X-Large
Material Microporous Non-woven Polyethylene Laminate
Color White
Seam Type Stitched, Taped
Certifications/Compliance (Europe) EN 374:2003, , EN 14605:2005 + A1:2009, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009, EN 1073-2:2002, EN 1149-5:2018, EN 14126:2003, EN ISO 13688:2013, DIN 32781:2010
Includes Hood and Boots
For Use With (Application) Product Inspection, Loading and Hauling, Fibre Glass/Resin Applications/Ceramic Fibres, Body Painting Including Surface Inspection, Cleaning of Plant and Machines, Maintaining Lab Furniture and Equipment
Weight (Metric) 63 gsm
Style W20T122