Learn More
Ansell AlphaTec™ 37-510 Food-Safe 11 mil. Nitrile Gloves with UltraGrip™ Technology
Product Code. 16805220
Click to view available options
Size:
7
8
9
10
11
12
Lightweight, reusable gloves specifically designed for food processing and handling. Raised diamond-pattern grip on the glove's surface enhances grip capabilities in wet and fatty conditions.
  • Reusable nitrile gloves designed specifically for food processing and handling
  • High performance nitrile maintains flexibility and dexterity, even in low temperature applications
  • Raised diamond-pattern enhances grip in wet and fatty conditions
  • Latex-free formulation presents no risk of Type 1 allergies
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Color Blue
Disposable Reusable
Hand Specific
Latex Free Yes
Material Nitrile
Size 9
Thickness (English) 11 mil
Thickness (Metric) 0.28 mm
EN388 Standard 2001A
EN 421 Yes
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Yes
Series 37-510