Learn More
Ansell AlphaTec™ 37-510 Food-Safe 11 mil. Nitrile Gloves with UltraGrip™ Technology
Product Code. 16815220
Click to view available options
Size:
7
8
9
10
11
12
Lightweight, reusable gloves specifically designed for food processing and handling. Raised diamond-pattern grip on the glove's surface enhances grip capabilities in wet and fatty conditions.
  • Reusable nitrile gloves designed specifically for food processing and handling
  • High performance nitrile maintains flexibility and dexterity, even in low temperature applications
  • Raised diamond-pattern enhances grip in wet and fatty conditions
  • Latex-free formulation presents no risk of Type 1 allergies
TRUSTED_SUSTAINABILITY
Specifications
ANSI Abrasion Rating ANSI 4
Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Color Blue
Disposable Reusable
Hand Specific
Latex Free Yes
Material Nitrile
Size 10
Thickness (English) 11 mil
Thickness (Metric) 0.28 mm
EN388 Standard 2001A
EN 421 Yes
EN ISO 374-1 CHEMICAL LETTER CODES JKL
EN ISO 374 - 5 Yes
Series 37-510