missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 38-560 Chemical Resistant Butyl Gloves - Gauntlet Cuff

Product Code. 16865220
Click to view available options
Size:
7
8
9
10
11
Unit Size:
10 pairs
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Product Code. 16865220

Brand: Ansell 38560090

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

Gloves are constructed of a thick butyl material to provide a high level of chemical resistance against agressive chemicals, without sacrificing comfort or dexterity.

  • High protection level against gases, particularly ozone
  • Extended gauntlet style cuff for additional protection
  • Beaded cuff for tear resistance and easy donning
  • Rough finish on the hand for safer handling
  • Designed for comfort with a natural, curved ergonomic shape
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Applications Industrial
Color Black
Latex Free Yes
Material Butyl Polymer
Product Type Butyl Gloves
Size 9
EN388 Standard 1020X
EN ISO 374-1 Type A
EN ISO 374-1 CHEMICAL LETTER CODES ABIKPT
EN ISO 374 - 5 Yes
Glove Cuff Style Gauntlet
Resistant To Solvents, and Strong Acids
Series 38-560
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.