missing translation for 'onlineSavingsMsg'
Learn More
Ansell™ Alphatec™ 55-101 Series Natural Rubber Latex Gloves Product Code.: 15630914
Add to basket

Ansell™ Alphatec™ 55-101 Series Natural Rubber Latex Gloves

Product Code. 15630914 Shop All Ansell Products
10
Click to view available options
Size:
8
9
10
This item is not returnable. View return policy

Product Code. 15630914

Brand: Ansell™ 55101100

Add to basket
Add to basket
This item is not returnable. View return policy

Provides effective protection for heavy duty physical environments. Ansell Alphatec™ 55-101 Series Natural Rubber Latex Gloves provides defense against tears and abrasions, cuts and punctures.

  • Chemical and liquid protection gloves
  • Highly resistant to most acids, alkalis, alcohols, aldehydes and ketones
  • Heavy weight thickness increases durability, augmenting chemical and physical resistance
  • Translucent and unlined for effective cleaning

Applications:

  • Biotechnologies
  • Blending of liquids
  • Chemical and Medical research
  • Chemical handling
  • Chemical industry
  • Chemicals mixing/handling/transferring/pouring/blending
  • Pharmaceuticals
  • Product handling - chemical/biological
  • Product protection applications
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 420:2003, A1:2009 and EN 388:2003, EN 374:2003
Color Natural
Exterior Finish Smooth
Material Rubber Latex
Product Type Chemical and Liquid Protection Gloves
Size 10
Thickness (Metric) 0.76 mm
Glove Cuff Style Beaded/Rolled
Packaging Type 10 pairs in black sealed bags per shipper carton
Port Size (Metric) 150 mm
Series 55-101
Length (Metric) 700 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Ansell™ Alphatec™ 55-101 Series Natural Rubber Latex Gloves >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.