missing translation for 'onlineSavingsMsg'
Learn More

Ansell™ AlphaTec™ 57-435 Series Nitrile Immersion Gloves

Product Code. 15599441
Click to view available options
Size:
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Provides confident grip performance with enhanced flexibility. Ansell Alphatec™ 58-435 Series Nitrile Immersion Gloves has optimal control when handling oily or wet items.

  • Chemical and liquid protection gloves
  • Features Ansell Grip Technology™ for optimal control when handling oily or wet items
  • Less force is required on slippery objects, reducing hand and arm fatigue
  • Offers improved flexibility and dexterity thanks to cotton-flocked lined nitrile shell and snug, second-skin tailoring that tightens the fit across the palm
  • Has 6-mil shell effective against hydrocarbon derivatives, alcohol bases, many solvents and esters
  • Offers superior snag, puncture and abrasion protection for long-term wear
  • Foldable gutter cuff helps prevent dripping onto the forearm, while an extra-long cuff extends protection to the upper forearm

Applications:

  • Chemical handling
  • Petrochemicals
  • Rubber and rubber additives
  • Handling fluid-coated metal parts
  • Industrial tool maintenance
  • Handling waste oils
  • Waste and water treatment
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance EN 374, EN 388:2003 4001
Color Green, Gray
Material Nitrile
Product Type Chemical and Liquid Protection Gloves
Size 7
Glove Cuff Style Gauntlet
Liner Material Cotton flocking
Packaging Type 12 pairs/polybag, 6 polybags/carton, 72 pairs/carton
Series 58-435
Length (Metric) 380 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.