Learn More
Ansell™ AlphaTec™ 58-530B Series Nitrile Immersion Gloves
Enables users to handle wet or oily objects with less grip force and more control. The patented Ansell Grip TechnologyTM ensures that the coating polymer does not penetrate the liner during manufacture significantly reducing the likelihood of chemical leakage onto the skin.
Brand: Ansell™ 58530B080
This item is not returnable.
View return policy
Description
- Nitrile polymer coating reduces chemical leakage onto skin
- For indoor applications
- Nylon liner
- Gauntlet cuff
- Sea green shell, anthracite grey dip color
Specifications
Nitrile | |
Green, Gray | |
8 | |
Gauntlet | |
Black Acrylic | |
Chemical resistance |
Nitrile | |
Immersion Gloves | |
58-530B | |
Acrylic | |
6 Pairs | |
58-530B |