missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ansell AlphaTec™ 38-628 Butyl/Viton Immersion Gloves
AlphaTec™ Gloves 38-628 offer a combination of butyl and viton layer of 0.7 mm for a maximum protection against hazardous chemicals. Provide the best resistance to the most aggressive chemicals without compromising dexterity or comfort. Offer superior protection against aliphatic, halogenated and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids.
Brand: Ansell 38628110
View more versions of this product
This item is not returnable.
View return policy
Description
- AlphaTec™ 38-628 offers superior protection against aliphatic, halogenated and aromatic hydrocarbons (benzene, toluene, xylene), as well as concentrated mineral acids
- The glove is very flexible and provides excellent dexterity
- Designed for comfort, with a natural, curved ergonomic shape and soft feel, offering easy donning and good grip
- The absence of latex proteins means there's no risk of latex allergies
Specifications
ANSI 2 | |
Industrial | |
27.6 mil | |
Yes | |
27.6 mil | |
11 | |
0.7 mm | |
No | |
ABCDFLNS | |
Smooth Finish | |
AlphaTec™ 38-628 | |
350 mm |
CE Number, EN ISO 374-1:2016, EN 388:2003, EN 388:2016, EN 388:2016 +A1:2018, EN 420:2003 + A1:2009, EN ISO 21420:2020, EN ISO 374-1:2016, EN ISO 374-5:2016, EN 374:2003, , EN ISO 374-5:2016, Category III, REACH Compliant | |
Black | |
0.7 mm | |
Viton, Butyl Polymer | |
0.7 mm | |
27.6 mil | |
2120A | |
Type A | |
Beaded/Rolled | |
Unlined | |
13.8 in. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction