missing translation for 'onlineSavingsMsg'
Learn More

Ansell AlphaTec™ 18 in. Solvex™ 37-185 Nitrile Gloves
GREENER_CHOICE

Heavy-duty, elbow length nitrile chemical resistant gloves suitable for a wide range of applications.

136.00 € - 138.00 €

Specifications

ANSI Abrasion Rating ANSI 4
Certifications/Compliance FDA 21 CFR 177.2600
Certifications/Compliance (Europe) CE 0493, EN 420:2003 + A1:2009, Category III
Chlorinated Yes
Color Green
View More Specs

Products 4
Product Code Brand Size Price Quantity & Availability  
Product Code Brand Size Price Quantity & Availability  
11741593
View Documents
Ansell
37185080
8
138.00 €
12 pairs
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
12445140
View Documents
Ansell
37185090
9
136.00 €
12 pairs
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
12015550
GREENER_CHOICE
missing translation for 'viewDocuments'
Ansell
37185100
10
136.00 €
12 pairs
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
12025550
missing translation for 'viewDocuments'
Ansell
37185110
11
136.00 €
12 pairs
Estimated Shipment: 17-01-2025
to see stock.
Add to basket
 
Description

Description

Heavy-duty, elbow length nitrile chemical resistant gloves suitable for a wide range of applications.

  • High performance nitrile compound
  • Combination of chemical resistance and strength ,for optimal results in wet or dry environments
  • Snag, puncture and abrasion protection compared with rubber or neoprene gloves
  • High levels of flexibility, comfort and dexterity
  • Suitable for heavy-duty cleaning applications
  • Complies with FDA food handling requirements (21 CFR 177.2600)
Specifications

Specifications

ANSI 4
CE 0493, EN 420:2003 + A1:2009, Category III
Green
Nitrile
0.55 mm
No
AGJKLPT
Chemical plants and transportation, chemical processing, Oil refining and Petrochemicals, cleaning and de-greasing, exposure to chemicals, Solvents, hazardous liquids, PVC cements or standing Water, janitorial and maintenance work
Sandpatch
ISO 18889
18 in.
FDA 21 CFR 177.2600
Yes
Yes
22 mil
EN 388:4102X, EN 388:4102
Type A
Bacteria, Fungi only
Gauntlet
Unlined
AlphaTec™ Solvex™ 37-185
45.7 cm
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.