Learn More
ALPL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11187-1-AP-150UL
 View more versions of this product
Product Code. 16878093
  • 410.00 € / 150µL
Explore more special offers
Description
Description
This antibody can bind the four mentioned alkaline phosphatases.
Alkaline phosphatase is a homodimeric protein enzyme (86 kDa), containing two zinc atoms crucial to its catalytic function per monomer, and it is optimally active at alkaline pH environments. The enzyme is found in prokaryotes and eukaryotes, with the same general function but in different structural forms suitable to organism′s environment. ALP plays a role in liver metabolism and development within the skeleton. In bacteria, ALP acts as a source of inorganic phosphate. In yeast, ALP can be a useful protein for studying membrane protein sorting.
Specifications
Specifications
ALPL
Polyclonal
Unconjugated
ALPL
Alkaline phosphatase, Alkaline phosphatase liver/bone/kidney isozyme, Alkaline phosphatase tissue nonspecific isozyme, ALPL, BLAP, Bone Alkaline Phosphatase, NBAP
Rabbit
Antigen Affinity Chromatography
RUO
248, 249
-20°C
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P05186, P09923
ALPI, ALPL
ALPL Fusion Protein Ag1664
150 μL