missing translation for 'onlineSavingsMsg'
Learn More

Alprostadil, Tocris Bioscience™

Product Code. 18782514
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Prostaglandin. Vasodilator and antiplatelet agent in vivo

Prostaglandin with some selectivity for EP3 and EP4 receptors (Ki values are 1.1, 2.1, 36, 10 and 33 nM for mouse EP3, EP4, EP1, EP2 and IP receptors respectively). Inhibits platelet aggregation and is a vasodilator in vivo.

Chemical Identifiers

CAS 745-65-3
Molecular Formula C20H34O5
Molecular Weight (g/mol) 354.487
InChI Key GMVPRGQOIOIIMI-DWKJAMRDSA-N
Synonym alprostadil, prostaglandin e1, pge1, muse, alprostadilum, prostavasin, topiglan, vasaprostan, femprox, edex
PubChem CID 5280723
ChEBI CHEBI:15544
IUPAC Name 7-[(1R,2R,3R)-3-hydroxy-2-[(E,3S)-3-hydroxyoct-1-enyl]-5-oxocyclopentyl]heptanoic acid
SMILES CCCCCC(C=CC1C(CC(=O)C1CCCCCCC(=O)O)O)O

Specifications

Quantity 50 mg
Formula Weight 354.49
Percent Purity >98%
Chemical Name or Material Alprostadil
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'