Learn More
Thermo Scientific™ ALPS 3000™ Automated Microplate Heat Sealer
GREENER_CHOICE
Seal in heat fast with this automated heat sealer, designed for both optimal robotic integration in high throughput labs and manual benchtop control.
Brand:  Thermo Scientific™ AB-3000
26695.50 EUR valid until 2024-12-15
Use promo code "21658" to get your promotional price.
Product Code. 11557369
  • 43595.00 € / Each
Explore more special offers
Description
Description
Quickly seal in heat with the Thermo Scientific™ ALPS 3000 Automated Microplate Heat Sealer. This compact sealer is designed for optimal robotic integration in high throughput labs, yet still allows for manual benchtop control.
Rapid Plate Sealing Action
  • Top-loaded seals are easily accessible
  • Seven available sealing options
  • Seals up to 10 plates per minute (six seconds/plate)
Fast, Easy to Use
  • Light (26 pounds) with a small footprint (165 × 405mm), takes up less precious bench space than comparable instruments
  • Available with a variety of available heat seals meet any laboratory application, including PCR, coulorimetric, fluorescence, long term storage, low temperature storage, piercing and resealing
  • On-board touch screen operation for benchtop use and and RS232 port with included ASCII programming parameters for robotic integration
Safe and Secure
  • Sensors detect the presence of sealing tape and plate
  • Air input allow for automatic system shutoff to eliminate product waste, system hold-ups and potential injury
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Plate Sealer
110/240 V
16.3 x 7.3 x 13.8 in. (414 x 185 x 350 mm)
13.8 in.
16.3 in.
1 Ea.
26.5 lb.
7.3 in.
50/60 Hz
ALPS 3000 Automated Microplate Heat Sealer
110/240 V, 50/60 Hz
350 mm
414 mm
200°C
12 kg
185 mm