Learn More
Macherey-Nagel™ Alugram Aluminum Sheet CEL 300 UV254
109.00 € - 170.00 €
Specifications
Type Aluminum Sheet
Fluorescent Indicator UV254
For Use With (Application) Partition Chromatography Of Polar Substances Such As Amino Acids, Carboxylic Acids Or Carbohydrates
Specific Surface 15,000 cm2/g
View More Specs
Products 2
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
Product Code Brand Dimensions (L x W) Price Quantity & Availability  
10338651
View Documents
Macherey-Nagel™
818156
20 x 20 cm
170.00 €
Pack of 25
In Stock
to see stock.
Add to basket
10516881
View Documents
Macherey-Nagel™
818157
5 x 2 cm
109.00 €
Pack of 50
In Stock
to see stock.
Add to basket
Specifications
Aluminum Sheet
UV254
Partition Chromatography Of Polar Substances Such As Amino Acids, Carboxylic Acids Or Carbohydrates
15,000 cm2/g
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Macherey-Nagel™ Alugram Aluminum Sheet CEL 300 UV254