missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ ALUGRAM™ Xtra SIL G Aluminum Sheets
Stable in almost all organic solvents and in purely aqueous eluents. Macherey-Nagel™ ALUGRAM™ Xtra SIL G Aluminum Sheets are unmodified standard silica layers on aluminum for use in TLC. Feature outstanding wettability and improved cutting properties.
27.20 € - 151.00 €
Specifications
Material | Alugram Aluminum Sheets with IUInmodified Silica 60 Layers |
---|---|
Particle Size | 5 to 17 μm |
Pore Size | 60 Å |
Type | Aluminum Sheet |
Pore Volume | 0.75 mL/g |
Product Code | Brand | Dimensions (L x W) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Code produit | Marque | Dimensions (L x W) | Prix | Quantité et disponibilité | ||||||
12876440
|
Macherey-Nagel™
818233 |
20 x 20 cm |
151.00 €
Pack of 25 |
In Stock
Log in to see stock. |
||||||
12730743
|
Macherey-Nagel™
818232 |
5 x 20 cm |
85.38 €
Pack of 50 |
In Stock
Log in to see stock. |
||||||
12720743
|
Macherey-Nagel™
818230.20 |
5 x 7.5 cm |
27.20 €
Pack of 20 |
In Stock
Log in to see stock. |
||||||
12750763
|
Macherey-Nagel™
818261 |
5 x 10 cm |
62.35 €
Pack of 50 |
In Stock
Log in to see stock. |
||||||
Description
ALUGRAM™ Xtra SIL G Aluminum Sheets feature unmodified, standard-grade silica 60 layers without a fluorescent indicator.
- Outstanding wettability for precise colorization results, even with 100% aqueous detection reagents
- Excellent separation efficiency and reproducibility from lot to lot
- Easy and reliable cutting due to an optimized binder system, no flaking of silica
- Stable in almost all organic solvents and resistant towards aggressive visualization reagents
- Highly polymeric product is also completely stable in purely aqueous eluents
Spécification
Alugram Aluminum Sheets with IUInmodified Silica 60 Layers | |
60 Å | |
0.75 mL/g | |
0.20 mm |
5 to 17 μm | |
Aluminum Sheet | |
∼500 m2/g |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Macherey-Nagel™ ALUGRAM™ Xtra SIL G Aluminum Sheets