missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254
Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Xtra SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.
Brand: Macherey-Nagel™ 818330.20
21.96 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Unmodified standard silica layers on aluminum for TLC
- Silica 60
- Outstanding wettability for precise colorization results, even with 100 % aqueous detection reagents
- Excellent separation efficiency and reproducibility from lot to lot
- Easy and reliable cutting due to an optimized binder system, no flaking of silica
- Binder: highly polymeric product, which is stable in almost all organic solvents and resistant towards aggressive visualization reagents; also completely stable in purely aqueous eluents
Specifications
5 to 17 μm | |
20 Pk. | |
UV254 | |
∼500 m2/g | |
5 x 7.5 cm |
60 Å | |
Aluminum Sheet | |
0.75 mL/g | |
0.20 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction