Learn More
Macherey-Nagel™ Alugram Aluminum Sheet SILGUR UV254
TLC aluminum sheets with optimized cuttability and wettability. Layer - unmodified silica gel (SiOH) with concentrating zone (kieselguhr, diatomaceous earth or diatomite).
Brand: Macherey-Nagel™ 818413
151.98 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
TLC aluminum sheets with optimized cuttability and wettability. Layer - unmodified silica gel (SiOH) with concentrating zone (kieselguhr, diatomaceous earth or diatomite).
Specifications
5 to 17 μm | |
25 Pk. | |
0.75 mL/g | |
0.20 mm |
60 Å | |
Aluminum Sheet | |
∼500 m2/g | |
20 x 20 cm |