missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alumina A, Activity: Super I, MP EcoChrom™, 63 to 200 μm, MP Biomedicals™
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
Description
- Adsorption capacity approximately twice as high as that of MP EcoChrom™ Alumina N, Activity I.
- Inital activity is kept in a very narrow range.
- Extremely constant deactivation behavior.
- Commonly used for purification processes requiring the highest precision and reproducibility.
Specifications
Specifications
Color | Pink to White |
Physical Form | Free-flowing Powder |
Quantity | 500 g |
IUPAC Name | dialuminium(3+) trioxidandiide |
Formula Weight | 101.96 |
Density | 3.94 g/cm3 (at 20°C) |
Chemical Name or Material | Alumina A, Activity: Super I |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'