missing translation for 'onlineSavingsMsg'
Learn More

Haldenwanger™ Alumina Tall Form Alsint Crucible

Alsint 99.7 crucibles are an excellent choice for use under high-wear conditions, in chemical technical and electrical engineering applications, and in high-temperature technology.

Brand:  Haldenwanger™ L ALSINTTIEGEL*C/0

Product Code. 10758361

  • 40.10 € / Each
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Applications include crystal growing, fusion processes, and annealing
  • Excellent for use with ball mill pots, milling balls, mouthpieces, and shaft-protection sleeves
  • High refractoriness (melting point above 2000°C)
  • High chemical resistance to a wide range of substances both at boiling and room temperature

Order Info

CapacityMetric

Specifications

Specifications

White
15 mL
38mm
Tall Form
Alumina
30/18mm
Alsint Crucible
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.