missing translation for 'onlineSavingsMsg'
Learn More

Aluminum, 99.9995%, (trace metal basis), shot, Thermo Scientific Chemicals

Product Code. 10078339
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Color Gray
CAS 7429-90-5
Melting Point 660.0°C
Boiling Point 2327.0°C
Density 2.7000g/mL
Assay Percent Range 99.9995%
Molecular Formula Al
MDL Number MFCD00134029
Quantity 25 g
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
SMILES [Al]
IUPAC Name aluminum
Molecular Weight (g/mol) 26.98
PubChem CID 5359268
ChEBI CHEBI:33629
Formula Weight 26.97
Percent Purity 99.9995%
Grade Trace Metal Basis
Packaging Glass bottle
Form Shot
Chemical Name or Material Aluminum
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'