missing translation for 'onlineSavingsMsg'
Learn More

Aluminum bromide, 98+%, extra pure, anhydrous, Thermo Scientific Chemicals

Product Code. 10433873
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-15-3
Molecular Formula AlBr3
Molecular Weight (g/mol) 266.69
MDL Number MFCD00003421
InChI Key PQLAYKMGZDUDLQ-UHFFFAOYSA-K
IUPAC Name aluminium(3+) tribromide
SMILES [Al+3].[Br-].[Br-].[Br-]

Specifications

Boiling Point 265.0°C
Melting Point 97.0°C
Color Gray or White
Physical Form Crystalline Solid
Quantity 25 g
Assay Percent Range 98% min. (Complexometry)
Linear Formula AlBr3
Fieser 01,22; 02,19; 03,7; 04,10; 05,10; 06,17; 09,11
Merck Index 15,328
Solubility Information Solubility in water: reacts. Other solubilities: soluble in alcohol, carbon disulfide, benzene,, toluene, ether, nitrobenzene and xylene
Formula Weight 266.7
Percent Purity ≥98%
Grade Extra Pure
Packaging Glass bottle
Chemical Name or Material Aluminum bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aluminium bromide, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.