Learn More
Aluminum fluoride trihydrate, 97%, extra pure, Thermo Scientific Chemicals
59.70 € - 207.03 €
Chemical Identifiers
CAS | 15098-87-0 |
---|---|
Molecular Formula | AlF3·3H2O |
Molecular Weight (g/mol) | 138.02 |
MDL Number | MFCD00149141 |
InChI Key | BKLZIAYVINRQEJ-UHFFFAOYSA-K |
Synonym | aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride |
PubChem CID | 22002992 |
IUPAC Name | aluminum;trifluoride;trihydrate |
SMILES | O.O.O.[F-].[F-].[F-].[Al+3] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10637182
|
Thermo Scientific Acros
216825000 |
500 g | Plastic bottle |
59.70 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10773724
|
Thermo Scientific Acros
216820025 |
2.5 kg | Plastic bottle |
207.03 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Lewis acidChemical Identifiers
15098-87-0 | |
138.02 | |
BKLZIAYVINRQEJ-UHFFFAOYSA-K | |
22002992 | |
O.O.O.[F-].[F-].[F-].[Al+3] |
AlF3·3H2O | |
MFCD00149141 | |
aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride | |
aluminum;trifluoride;trihydrate |
Specifications
15098-87-0 | |
Powder | |
AlF3·3H2O | |
MFCD00149141 | |
aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride | |
BKLZIAYVINRQEJ-UHFFFAOYSA-K | |
aluminum;trifluoride;trihydrate | |
22002992 | |
97% | |
Aluminum fluoride trihydrate, 97% |
White | |
96% min | |
AlF3·3H2O | |
10, 333 | |
Solubility in water: slightly soluble | |
O.O.O.[F-].[F-].[F-].[Al+3] | |
138.02 | |
138.02 | |
Extra Pure |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
- Aluminium fluoride trihydrate
RUO – Research Use Only