missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Nitrate Nonahydrate, 98+%, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
250 g
500 g
1 kg
5 kg
Specifications
Specifications
Melting Point | 73°C |
pH | 2.5 to 3.5 |
Physical Form | Solid |
Quantity | 500 g |
Formula Weight | 375.13g/mol |
Packaging | Plastic powder jar |
Safety and Handling
Product Identifier
- Aluminium nitrate nonahydrate
- Warning
- Oxidising solid Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- MIXTURE LIST-Contains : Aluminium nitrate nonahydrate, Aluminumnitrate
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction