missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, for chromatography, basic, Brockmann I, 40-300μm, 60A, Thermo Scientific Chemicals

Product Code. 10730701
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Aluminum oxide, Al2O3, CAS Number-1344-28-1, abramant, abramax, abrarex, abrasit, aloxite, alpha-alumina, Aluminum oxide, Aluminum oxide, alundum, fasertonerde, 1kg, Na2O <0.4%, 2977.0 deg.C, White, 3.9700g/mL, 215-691-6, 300ppm max.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For column chromatography
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2977.0°C
Melting Point 2030.0°C
Color White
pH 9 to 10 (10% aq. suspension)
Physical Form Powder
Quantity 1 kg
Linear Formula Al2O3
Fieser 01,19; 02,17; 03,6; 04,8; 06,16; 07,5; 08,9; 09,8; 10,8; 11,22; 13,14; 15,9; 16,9
Merck Index 15, 350
Solubility Information Solubility in water: insoluble. Other solubilities: slightly sol. in alkaline sol.(forming hydroxides), practically insoluble in non-polar organic solvent, difficultly soluble in mineral acids
IUPAC Name dialuminium(3+) trioxidandiide
Formula Weight 101.96
Grade Chromatography
Packaging Plastic Bottle
Specific Gravity 3.97
Density 3.9700g/mL
Chemical Name or Material Aluminum oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.