missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide, Extra Pure, SLR, Calcined, Fisher Chemical™

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
IUPAC Name dialuminium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Description

Description

Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
[O--].[O--].[O--].[Al+3].[Al+3]
Al2O3
MFCD00003424
dialuminium(3+) trioxidandiide

Specifications

2980°C
1344-28-1
Solid
MFCD00003424
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
Plastic powder jar
2000°C
White
Al2O3
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminium(3+) trioxidandiide
101.96g/mol
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.