Learn More
Aluminum potassium sulfate dodecahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
32.20 € - 107.00 €
Chemical Identifiers
CAS | 7784-24-9 |
---|---|
Molecular Formula | AlKO8S2·12H2O |
Molecular Weight (g/mol) | 474.39 |
MDL Number | MFCD00149143 |
InChI Key | GNHOJBNSNUXZQA-UHFFFAOYSA-J |
Synonym | kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate |
PubChem CID | 62667 |
ChEBI | CHEBI:86465 |
IUPAC Name | aluminum;potassium;disulfate;dodecahydrate |
SMILES | O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10120362
|
Thermo Scientific Acros
217485000 |
500 g | Plastic bottle |
32.20 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10701111
|
Thermo Scientific Acros
217480010 |
1 kg | Plastic bottle |
53.40 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10509243
|
Thermo Scientific Acros
217480025 |
2.5 kg | Plastic bottle |
107.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7784-24-9 | |
474.39 | |
GNHOJBNSNUXZQA-UHFFFAOYSA-J | |
62667 | |
aluminum;potassium;disulfate;dodecahydrate |
AlKO8S2·12H2O | |
MFCD00149143 | |
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate | |
CHEBI:86465 | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+] |
Specifications
92.0°C | |
White | |
Crystals | |
AlKO8S2·12H2O | |
MFCD00149143 | |
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate | |
GNHOJBNSNUXZQA-UHFFFAOYSA-J | |
aluminum;potassium;disulfate;dodecahydrate | |
62667 | |
474.39 | |
Analytical |
7784-24-9 | |
3 to 3.5 (10% soln. at 20°C) | |
99.5% min. (Complexometry) | |
AlK(SO4)2·12H2O | |
15, 354 | |
Solubility in water: 140g/L (20°C). Other solubilities: freely soluble in glycerol, insoluble in alcohol | |
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+] | |
474.39 | |
CHEBI:86465 | |
99.5% | |
Aluminum potassium sulfate dodecahydrate, p.a. |
Safety and Handling
GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Warning
RUO – Research Use Only